Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MIIP
Entrez Gene ID
60672
Gene Name
migration and invasion inhibitory protein
Gene Aliases
IIP45
Chromosome Location
Chr.1: 12019242 - 12032049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021933.3 NP_068752.2 5 - 6 1055 97
XM_005263487.3 XP_005263544.1 5 - 6 884 97
XM_011541895.1 XP_011540197.1 5 - 6 886 97
XM_011541896.1 XP_011540198.1 5 - 6 779 97
GenBank mRNA AK024020.1 - 5 - 6 774 97
AK095928.1 - 2 - 3 169 97
BC008068.2 - 5 - 6 764 97
FJ618905.1 - 5 - 6 1055 97

View Full Product Details