The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012128.3 | NP_036260.2 | 11 - 12 | 1993 | 141 |
NR_024602.1 | 10 - 11 | 1884 | 141 | ||
XM_011541015.2 | XP_011539317.1 | 11 - 12 | 1812 | 141 | |
GenBank mRNA | AF127035.1 | - | 11 - 12 | 1977 | 141 |
AK000072.1 | - | 11 - 12 | 1991 | 141 | |
AK000138.1 | - | 4 - 5 | 670 | 141 | |
AK307785.1 | - | 10 - 11 | 1829 | 141 | |
AK307898.1 | - | 9 - 10 | 1598 | 141 | |
AK314430.1 | - | 11 - 12 | 1989 | 141 | |
AY358470.1 | - | 11 - 12 | 1973 | 141 | |
BC113687.1 | - | 11 - 12 | 1988 | 141 | |
BC113689.1 | - | 11 - 12 | 1987 | 141 |