The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012128.3 | NP_036260.2 | 12 - 13 | 2164 | 84 |
NR_024602.1 | 11 - 12 | 2055 | 84 | ||
XM_011541015.2 | XP_011539317.1 | 12 - 13 | 1983 | 84 | |
GenBank mRNA | AF127035.1 | - | 12 - 13 | 2148 | 84 |
AK000072.1 | - | 12 - 13 | 2162 | 84 | |
AK000138.1 | - | 5 - 6 | 841 | 84 | |
AK307785.1 | - | 11 - 12 | 2000 | 84 | |
AK307898.1 | - | 10 - 11 | 1769 | 84 | |
AK314430.1 | - | 12 - 13 | 2160 | 84 | |
AY358470.1 | - | 12 - 13 | 2144 | 84 | |
BC113687.1 | - | 12 - 13 | 2159 | 84 | |
BC113689.1 | - | 12 - 13 | 2158 | 84 |