Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLCA1
Entrez Gene ID
1179
Gene Name
chloride channel accessory 1
Gene Aliases
CACC, CACC1, CLCRG1, CaCC-1, GOB5, hCLCA1, hCaCC-1
Chromosome Location
Chr.1: 86468843 - 86500294 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285.3 NP_001276.2 1 - 2 294 144
GenBank mRNA AF039400.1 - 2 - 3 516 144
AF127036.1 - 1 - 2 169 144
AK291999.1 - 1 - 2 210 144
AK314375.1 - 1 - 2 212 144
DA428844.1 - 1 - 2 294 144

View Full Product Details