The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015394.4 | NP_056209.2 | 3 - 4 | 352 | 66 |
GenBank mRNA | AK314784.1 | - | 3 - 4 | 613 | 66 |
AL110188.1 | - | 1 - 2 | 69 | 66 | |
BC024182.2 | - | 3 - 4 | 353 | 66 | |
X52332.1 | - | 2 - 3 | 236 | 66 |