The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014448.3 | NP_055263.2 | 11 - 12 | 1870 | 69 |
XM_017001049.1 | XP_016856538.1 | 11 - 12 | 2471 | 69 | |
XM_017001050.1 | XP_016856539.1 | 11 - 12 | 1676 | 69 | |
XM_017001051.1 | XP_016856540.1 | 11 - 12 | 1881 | 69 | |
GenBank mRNA | AK095984.1 | - | 11 - 12 | 1777 | 69 |
AK296150.1 | - | 6 - 7 | 768 | 69 | |
BC002681.1 | - | 10 - 11 | 1157 | 69 | |
BC051838.1 | - | 9 - 10 | 1177 | 69 | |
BT007270.1 | - | 6 - 7 | 758 | 69 | |
D89016.1 | - | 8 - 9 | 1185 | 69 |