The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004288.4 | NP_004279.3 | 2 - 3 | 296 | 145 |
GenBank mRNA | AF068836.1 | - | 2 - 3 | 257 | 145 |
AK312753.1 | - | 2 - 3 | 293 | 145 | |
BC036449.2 | - | 2 - 3 | 254 | 145 | |
CD642163.1 | - | 2 - 3 | 349 | 145 | |
DB219691.1 | - | 2 - 3 | 297 | 145 | |
L06633.1 | - | 2 - 3 | 210 | 145 |