The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004288.4 | NP_004279.3 | 3 - 4 | 354 | 105 |
XM_017005386.1 | XP_016860875.1 | 3 - 4 | 545 | 105 | |
GenBank mRNA | AF068836.1 | - | 3 - 4 | 315 | 105 |
AK301544.1 | - | 6 - 7 | 447 | 105 | |
AK312753.1 | - | 3 - 4 | 351 | 105 | |
BC036449.2 | - | 3 - 4 | 312 | 105 | |
CD642163.1 | - | 3 - 4 | 407 | 105 | |
DB219691.1 | - | 3 - 4 | 355 | 105 | |
L06633.1 | - | 3 - 4 | 268 | 105 |