The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004288.4 | NP_004279.3 | 4 - 5 | 457 | 75 |
XM_017005386.1 | XP_016860875.1 | 4 - 5 | 648 | 75 | |
GenBank mRNA | AF068836.1 | - | 4 - 5 | 418 | 75 |
AK301544.1 | - | 7 - 8 | 550 | 75 | |
AK312753.1 | - | 4 - 5 | 454 | 75 | |
BC036449.2 | - | 4 - 5 | 415 | 75 | |
CD642163.1 | - | 4 - 5 | 510 | 75 | |
DB219691.1 | - | 4 - 5 | 458 | 75 | |
L06633.1 | - | 4 - 5 | 371 | 75 |