The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004288.4 | NP_004279.3 | 5 - 6 | 545 | 89 |
XM_017005386.1 | XP_016860875.1 | 5 - 6 | 736 | 89 | |
GenBank mRNA | AF068836.1 | - | 5 - 6 | 506 | 89 |
AK301544.1 | - | 8 - 9 | 638 | 89 | |
BC036449.2 | - | 5 - 6 | 503 | 89 | |
L06633.1 | - | 5 - 6 | 459 | 89 |