The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001287218.1 | NP_001274147.1 | 1 - 2 | 275 | 110 |
NM_001287220.1 | NP_001274149.1 | 1 - 2 | 275 | 110 | |
NM_012424.4 | NP_036556.2 | 1 - 2 | 275 | 110 | |
XM_005273095.1 | XP_005273152.1 | 1 - 2 | 262 | 110 | |
XM_006711272.1 | XP_006711335.1 | 1 - 2 | 262 | 110 | |
XM_017001025.1 | XP_016856514.1 | 1 - 2 | 248 | 110 | |
XM_017001026.1 | XP_016856515.1 | 1 - 2 | 250 | 110 | |
XM_017001032.1 | XP_016856521.1 | 1 - 2 | 250 | 110 | |
GenBank mRNA | AF477978.1 | - | 1 - 2 | 252 | 110 |
AK122989.1 | - | 1 - 2 | 262 | 110 | |
BC104769.1 | - | 1 - 2 | 189 | 110 | |
BC143473.1 | - | 1 - 2 | 189 | 110 | |
DB060692.1 | - | 1 - 2 | 263 | 110 |