The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138381.3 | NP_612390.1 | 2 - 3 | 499 | 131 |
XM_006713406.2 | XP_006713469.1 | 2 - 3 | 448 | 131 | |
XM_017007486.1 | XP_016862975.1 | 2 - 3 | 495 | 131 | |
XM_017007487.1 | XP_016862976.1 | 2 - 3 | 495 | 131 | |
XM_017007488.1 | XP_016862977.1 | 2 - 3 | 495 | 131 | |
XM_017007489.1 | XP_016862978.1 | 2 - 3 | 495 | 131 | |
XM_017007490.1 | XP_016862979.1 | 2 - 3 | 495 | 131 | |
XM_017007491.1 | XP_016862980.1 | 2 - 3 | 495 | 131 | |
XM_017007492.1 | XP_016862981.1 | 2 - 3 | 234 | 131 | |
XM_017007493.1 | XP_016862982.1 | 2 - 3 | 448 | 131 | |
XM_017007494.1 | XP_016862983.1 | 2 - 3 | 448 | 131 | |
XM_017007495.1 | XP_016862984.1 | 2 - 3 | 445 | 131 | |
XM_017007496.1 | XP_016862985.1 | 2 - 3 | 445 | 131 | |
GenBank mRNA | AK307258.1 | - | 2 - 3 | 426 | 131 |
AL583453.3 | - | 2 - 3 | 458 | 131 | |
BC008322.1 | - | 2 - 3 | 426 | 131 | |
DQ364598.1 | - | 2 - 3 | 457 | 131 |