The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005285.3 | NP_005276.2 | 1 - 1 | 743 | 130 |
GenBank mRNA | BC069117.1 | - | 1 - 1 | 743 | 130 |
BC107101.2 | - | 1 - 1 | 743 | 130 |