The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198333.2 | NP_938147.1 | 1 - 2 | 166 | 106 |
GenBank mRNA | AK313218.1 | - | 1 - 2 | 107 | 106 |
CD639881.1 | - | 1 - 2 | 147 | 106 |