The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024758.4 | NP_079034.3 | 3 - 4 | 673 | 93 |
GenBank mRNA | AK130034.1 | - | 3 - 4 | 469 | 93 |
AK225366.1 | - | 3 - 4 | 586 | 93 | |
AY057097.1 | - | 3 - 4 | 573 | 93 | |
BC005090.1 | - | 3 - 4 | 673 | 93 | |
DN996958.1 | - | 3 - 4 | 663 | 93 |