Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
Chromosome Location
Chr.1: 15681506 - 15734769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015164.2 NP_055979.2 15 - 16 2623 61
XM_005245790.3 XP_005245847.1 14 - 15 2758 61
XM_005245791.4 XP_005245848.1 10 - 11 1891 61
XM_017000757.1 XP_016856246.1 15 - 16 2530 61
XM_017000758.1 XP_016856247.1 14 - 15 2470 61
GenBank mRNA AB020649.1 - 15 - 16 2399 61
BC008002.1 - 7 - 8 660 61
BC016488.1 - 7 - 8 1305 61
BC030545.1 - 6 - 7 603 61
BC040441.1 - 15 - 16 2626 61
BC042103.1 - 7 - 8 953 61
BC068599.1 - 7 - 8 978 61

View Full Product Details