The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015164.2 | NP_055979.2 | 6 - 7 | 876 | 69 |
XM_017000757.1 | XP_016856246.1 | 6 - 7 | 783 | 69 | |
GenBank mRNA | AB020649.1 | - | 6 - 7 | 652 | 69 |
BC040441.1 | - | 6 - 7 | 879 | 69 |