Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLEKHM2
Entrez Gene ID
23207
Gene Name
pleckstrin homology and RUN domain containing M2
Gene Aliases
SKIP
Chromosome Location
Chr.1: 15681506 - 15734769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015164.2 NP_055979.2 9 - 10 1988 60
XM_005245790.3 XP_005245847.1 8 - 9 2123 60
XM_005245791.4 XP_005245848.1 4 - 5 1256 60
XM_017000757.1 XP_016856246.1 9 - 10 1895 60
XM_017000758.1 XP_016856247.1 8 - 9 1835 60
GenBank mRNA AB020649.1 - 9 - 10 1764 60
BC016488.1 - 1 - 2 670 60
BC040441.1 - 9 - 10 1991 60
BC042103.1 - 1 - 2 318 60
BC068599.1 - 1 - 2 343 60

View Full Product Details