Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM87B
Entrez Gene ID
84910
Gene Name
transmembrane protein 87B
Gene Aliases
-
Chromosome Location
Chr.2: 112055223 - 112119318 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032824.2 NP_116213.1 3 - 4 685 148
XM_005263827.2 XP_005263884.1 3 - 4 642 148
XM_006712807.2 XP_006712870.1 3 - 4 642 148
XM_017005121.1 XP_016860610.1 3 - 4 188 148
GenBank mRNA AK027587.1 - 3 - 4 670 148
AK290294.1 - 3 - 4 331 148
AY429534.1 - 3 - 4 188 148
AY429535.1 - 3 - 4 188 148
BC115373.1 - 3 - 4 489 148

View Full Product Details