The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024508.4 | NP_078784.2 | 2 - 2 | 766 | 87 |
GenBank mRNA | BC071956.1 | - | 1 - 1 | 231 | 87 |