Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WTIP
Entrez Gene ID
126374
Gene Name
Wilms tumor 1 interacting protein
Gene Aliases
-
Chromosome Location
Chr.19: 34481935 - 34509147 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080436.1 NP_001073905.1 1 - 2 668 71
XM_006723014.3 XP_006723077.1 1 - 2 701 71
XM_011526452.2 XP_011524754.1 1 - 2 708 71

View Full Product Details