The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080436.1 | NP_001073905.1 | 6 - 7 | 1082 | 82 |
XM_006723014.3 | XP_006723077.1 | 6 - 7 | 1115 | 82 | |
XM_011526452.2 | XP_011524754.1 | 6 - 7 | 1122 | 82 | |
XM_011526453.2 | XP_011524755.1 | 6 - 7 | 516 | 82 | |
GenBank mRNA | AK130059.1 | - | 5 - 6 | 420 | 82 |