The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000175.3 | NP_000166.2 | 14 - 15 | 1408 | 69 |
NM_001184722.1 | NP_001171651.1 | 14 - 15 | 1471 | 69 | |
NM_001289789.1 | NP_001276718.1 | 15 - 16 | 1555 | 69 | |
NM_001289790.1 | NP_001276719.1 | 13 - 14 | 1324 | 69 | |
XM_005258764.2 | XP_005258821.1 | 15 - 16 | 1374 | 69 | |
XM_006723148.2 | XP_006723211.1 | 15 - 16 | 1325 | 69 | |
XM_011526754.2 | XP_011525056.1 | 16 - 17 | 1790 | 69 | |
GenBank mRNA | AK025421.1 | - | 3 - 4 | 1006 | 69 |
AK129884.1 | - | 15 - 16 | 1339 | 69 | |
AK130102.1 | - | 13 - 14 | 1103 | 69 | |
AK293446.1 | - | 13 - 14 | 1200 | 69 | |
AK294396.1 | - | 14 - 15 | 1419 | 69 | |
AK301103.1 | - | 14 - 15 | 1212 | 69 | |
BC004982.1 | - | 14 - 15 | 1276 | 69 | |
K03515.1 | - | 14 - 15 | 1283 | 69 | |
X16539.1 | - | 14 - 15 | 1274 | 69 |