Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNDC9
Entrez Gene ID
10190
Gene Name
thioredoxin domain containing 9
Gene Aliases
APACD, PHLP3
Chromosome Location
Chr.2: 99306184 - 99336397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005783.3 NP_005774.2 2 - 3 380 126
XM_017003147.1 XP_016858636.1 2 - 3 339 126
GenBank mRNA AB006679.1 - 2 - 3 318 126
AB451403.1 - 1 - 2 189 126
AK222616.1 - 2 - 3 306 126
AK301722.1 - 3 - 4 350 126
AK313777.1 - 2 - 3 300 126
BC005968.1 - 2 - 3 316 126
BC022864.1 - 2 - 3 320 126
BC024223.2 - 2 - 3 320 126
BC070183.1 - 2 - 3 314 126
CR456935.1 - 1 - 2 189 126

View Full Product Details