The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321395.1 | NP_001308324.1 | 10 - 11 | 1061 | 69 |
NM_152781.3 | NP_689994.2 | 10 - 11 | 1181 | 69 | |
NR_135630.1 | 10 - 11 | 1225 | 69 | ||
XM_005278233.2 | XP_005278290.1 | 11 - 12 | 1140 | 69 | |
XM_005278235.1 | XP_005278292.1 | 11 - 12 | 1025 | 69 | |
XM_006721802.3 | XP_006721865.1 | 7 - 8 | 663 | 69 | |
XM_011524586.2 | XP_011522888.1 | 10 - 11 | 1124 | 69 | |
XM_017024424.1 | XP_016879913.1 | 10 - 11 | 1161 | 69 | |
GenBank mRNA | AK057392.1 | - | 10 - 11 | 1181 | 69 |
AK301774.1 | - | 11 - 12 | 1025 | 69 | |
AK301884.1 | - | 11 - 12 | 1145 | 69 | |
AK301943.1 | - | 10 - 11 | 1041 | 69 | |
BC033734.1 | - | 9 - 10 | 1815 | 69 | |
BC132681.1 | - | 10 - 11 | 1117 | 69 | |
BC144011.1 | - | 10 - 11 | 1161 | 69 |