Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
Chromosome Location
Chr.17: 35854946 - 35868891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321395.1 NP_001308324.1 12 - 13 1253 63
NM_152781.3 NP_689994.2 12 - 13 1373 63
NR_135630.1 12 - 13 1417 63
XM_005278233.2 XP_005278290.1 13 - 14 1332 63
XM_005278235.1 XP_005278292.1 13 - 14 1217 63
XM_006721802.3 XP_006721865.1 9 - 10 855 63
XM_011524586.2 XP_011522888.1 12 - 13 1316 63
XM_017024424.1 XP_016879913.1 12 - 13 1353 63
GenBank mRNA AK057392.1 - 12 - 13 1373 63
AK301943.1 - 12 - 13 1233 63
BC033734.1 - 11 - 12 2007 63
BC132681.1 - 12 - 13 1309 63
BC144011.1 - 12 - 13 1353 63

View Full Product Details