Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
Chromosome Location
Chr.17: 35854946 - 35868891 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321395.1 NP_001308324.1 8 - 9 791 60
NM_152781.3 NP_689994.2 8 - 9 911 60
NR_135630.1 8 - 9 104
XM_005278233.2 XP_005278290.1 9 - 10 870 60
XM_005278235.1 XP_005278292.1 9 - 10 755 60
XM_006721802.3 XP_006721865.1 5 - 6 393 60
XM_011524586.2 XP_011522888.1 8 - 9 854 60
XM_017024424.1 XP_016879913.1 8 - 9 891 60
GenBank mRNA AK057392.1 - 8 - 9 911 60
AK301774.1 - 9 - 10 755 60
AK301884.1 - 9 - 10 875 60
AK301943.1 - 8 - 9 771 60
BC033734.1 - 7 - 8 1545 60
BC132681.1 - 8 - 9 847 60
BC144011.1 - 8 - 9 104

View Full Product Details