Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GOLGA6A
- Entrez Gene ID
-
342096
- Gene Name
- golgin A6 family member A
- Gene Aliases
-
GLP, GOLGA6
- Chromosome Location
-
Chr.15: 74069857 - 74082550 on Build GRCh38
- Gene Symbol
- GOLGA6B
- Entrez Gene ID
-
55889
- Gene Name
- golgin A6 family member B
- Gene Aliases
-
GOLGA, GOLGA6D
- Chromosome Location
-
Chr.15: 72638291 - 72669604 on Build GRCh38
- Gene Symbol
- GOLGA6C
- Entrez Gene ID
-
653641
- Gene Name
- golgin A6 family member C
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 75258404 - 75273455 on Build GRCh38
- Gene Symbol
- LOC441728
- Entrez Gene ID
-
441728
- Gene Name
- golgin-like pseudogene
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 75775341 - 75789716 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK302879.1 |
- |
6 - 7 |
504 |
85 |
View Full Product Details