Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNH8
Entrez Gene ID
131096
Gene Name
potassium voltage-gated channel subfamily H member 8
Gene Aliases
ELK, ELK1, Kv12.1, elk3
Chromosome Location
Chr.3: 19148454 - 19535643 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144633.2 NP_653234.2 3 - 4 642 100
XM_017005698.1 XP_016861187.1 3 - 4 713 100
XM_017005699.1 XP_016861188.1 2 - 3 379 100
XM_017005700.1 XP_016861189.1 3 - 4 713 100
XM_017005701.1 XP_016861190.1 3 - 4 700 100
XM_017005702.1 XP_016861191.1 2 - 3 379 100
GenBank mRNA AB209053.1 - 3 - 4 633 100
AK295633.1 - 3 - 4 689 100
AK309034.1 - 3 - 4 618 100
AY053503.1 - 3 - 4 642 100

View Full Product Details