The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000880.3 | NP_000871.1 | 1 - 2 | 605 | 108 |
NM_001199886.1 | NP_001186815.1 | 1 - 2 | 605 | 108 | |
NM_001199887.1 | NP_001186816.1 | 1 - 2 | 605 | 108 | |
NM_001199888.1 | NP_001186817.1 | 1 - 2 | 605 | 108 | |
XM_011517522.2 | XP_011515824.1 | 1 - 2 | 545 | 108 | |
XM_011517523.2 | XP_011515825.1 | 1 - 2 | 545 | 108 | |
XM_017013397.1 | XP_016868886.1 | 1 - 2 | 545 | 108 | |
GenBank mRNA | AK226000.1 | - | 1 - 2 | 546 | 108 |
AK291538.1 | - | 1 - 2 | 605 | 108 | |
BC047698.1 | - | 1 - 2 | 593 | 108 | |
J04156.1 | - | 1 - 2 | 388 | 108 |