The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318889.1 | NP_001305818.1 | 2 - 3 | 576 | 138 |
NM_005816.4 | NP_005807.1 | 2 - 3 | 576 | 138 | |
NM_198196.2 | NP_937839.1 | 2 - 3 | 576 | 138 | |
NR_134917.1 | 2 - 3 | 576 | 138 | ||
XM_005247063.3 | XP_005247120.1 | 2 - 3 | 537 | 138 | |
XM_006713469.3 | XP_006713532.1 | 2 - 3 | 537 | 138 | |
XM_006713470.3 | XP_006713533.1 | 2 - 3 | 537 | 138 | |
XM_017005521.1 | XP_016861010.1 | 2 - 3 | 537 | 138 | |
XM_017005522.1 | XP_016861011.1 | 2 - 3 | 537 | 138 | |
GenBank mRNA | AK290719.1 | - | 2 - 3 | 462 | 138 |
AK315237.1 | - | 2 - 3 | 458 | 138 | |
AL833681.1 | - | 2 - 3 | 423 | 138 | |
BC020749.1 | - | 2 - 3 | 507 | 138 | |
BM561433.1 | - | 2 - 3 | 576 | 138 |