The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318889.1 | NP_001305818.1 | 4 - 5 | 922 | 74 |
NM_005816.4 | NP_005807.1 | 4 - 5 | 922 | 74 | |
NM_198196.2 | NP_937839.1 | 5 - 6 | 970 | 74 | |
NR_134917.1 | 4 - 5 | 922 | 74 | ||
XM_005247063.3 | XP_005247120.1 | 5 - 6 | 931 | 74 | |
XM_006713469.3 | XP_006713532.1 | 5 - 6 | 931 | 74 | |
XM_006713470.3 | XP_006713533.1 | 4 - 5 | 883 | 74 | |
XM_017005521.1 | XP_016861010.1 | 5 - 6 | 931 | 74 | |
GenBank mRNA | AK290719.1 | - | 4 - 5 | 808 | 74 |
AK315237.1 | - | 4 - 5 | 804 | 74 | |
AL833681.1 | - | 5 - 6 | 817 | 74 | |
BC020749.1 | - | 4 - 5 | 853 | 74 |