The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015976.4 | NP_057060.2 | 3 - 4 | 525 | 71 |
NR_033716.1 | 4 - 5 | 645 | 71 | ||
XM_006710677.2 | XP_006710740.1 | 3 - 4 | 542 | 71 | |
XM_006710678.2 | XP_006710741.1 | 4 - 5 | 641 | 71 | |
XM_011541564.2 | XP_011539866.1 | 4 - 5 | 581 | 71 | |
XM_017001425.1 | XP_016856914.1 | 3 - 4 | 503 | 71 | |
XM_017001426.1 | XP_016856915.1 | 4 - 5 | 640 | 71 | |
XM_017001427.1 | XP_016856916.1 | 4 - 5 | 915 | 71 | |
XM_017001428.1 | XP_016856917.1 | 3 - 4 | 390 | 71 | |
GenBank mRNA | AF121857.1 | - | 3 - 4 | 368 | 71 |
AK025031.1 | - | 3 - 4 | 482 | 71 | |
AK223229.1 | - | 3 - 4 | 384 | 71 | |
AK293018.1 | - | 3 - 4 | 453 | 71 | |
AL548028.3 | - | 3 - 4 | 526 | 71 | |
BC010349.1 | - | 3 - 4 | 339 | 71 | |
BC018105.1 | - | 4 - 5 | 463 | 71 | |
BI856196.1 | - | 3 - 4 | 546 | 71 | |
BT006747.1 | - | 2 - 3 | 281 | 71 |