The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038618.2 | NP_001033707.1 | 8 - 9 | 1129 | 75 |
NM_001083608.1 | NP_001077077.1 | 7 - 8 | 827 | 75 | |
NM_012336.3 | NP_036468.1 | 8 - 9 | 971 | 75 | |
NM_031968.2 | NP_114174.1 | 9 - 10 | 1109 | 75 | |
XM_005256340.3 | XP_005256397.1 | 8 - 9 | 1137 | 75 | |
XM_006722278.3 | XP_006722341.1 | 7 - 8 | 924 | 75 | |
XM_011523541.2 | XP_011521843.1 | 8 - 9 | 1061 | 75 | |
XM_011523543.2 | XP_011521845.1 | 9 - 10 | 974 | 75 | |
XM_017024449.1 | XP_016879938.1 | 9 - 10 | 1721 | 75 | |
XM_017024450.1 | XP_016879939.1 | 9 - 10 | 1211 | 75 | |
GenBank mRNA | AF128406.1 | - | 8 - 9 | 922 | 75 |
AK000929.1 | - | 8 - 9 | 924 | 75 | |
AK056966.1 | - | 8 - 9 | 1104 | 75 | |
BC000438.2 | - | 9 - 10 | 1076 | 75 | |
BC016440.1 | - | 7 - 8 | 714 | 75 |