The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000965.4 | NP_000956.2 | 6 - 7 | 1461 | 64 |
NM_001290216.2 | NP_001277145.1 | 9 - 10 | 1504 | 64 | |
NM_001290217.1 | NP_001277146.1 | 6 - 7 | 919 | 64 | |
NM_001290266.1 | NP_001277195.1 | 6 - 7 | 1104 | 64 | |
NM_001290276.1 | NP_001277205.1 | 6 - 7 | 1461 | 64 | |
NM_001290277.1 | NP_001277206.1 | 6 - 7 | 1323 | 64 | |
NM_001290300.1 | NP_001277229.1 | 6 - 7 | 925 | 64 | |
NM_016152.3 | NP_057236.1 | 6 - 7 | 1104 | 64 | |
NR_110892.1 | 5 - 6 | 1300 | 64 | ||
GenBank mRNA | AB307688.1 | - | 6 - 7 | 992 | 64 |
AF157483.1 | - | 6 - 7 | 1103 | 64 | |
AK299756.1 | - | 6 - 7 | 1079 | 64 | |
AY501390.1 | - | 6 - 7 | 893 | 64 | |
BC050415.2 | - | 4 - 5 | 687 | 64 | |
BC060794.1 | - | 6 - 7 | 1143 | 64 | |
CD014021.1 | - | 5 - 6 | 847 | 64 | |
CD014022.1 | - | 6 - 7 | 870 | 64 | |
CR536558.1 | - | 4 - 5 | 656 | 64 | |
HQ692863.1 | - | 4 - 5 | 656 | 64 | |
HQ692864.1 | - | 6 - 7 | 992 | 64 | |
X07282.1 | - | 6 - 7 | 1335 | 64 | |
Y00291.1 | - | 6 - 7 | 1313 | 64 |