The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308164.1 | NP_001295093.1 | 2 - 3 | 336 | 66 |
NM_001308166.1 | NP_001295095.1 | 2 - 3 | 336 | 66 | |
NM_024996.5 | NP_079272.4 | 2 - 3 | 336 | 66 | |
XM_006713795.1 | XP_006713858.1 | 2 - 3 | 367 | 66 | |
GenBank mRNA | AF309777.1 | - | 2 - 3 | 336 | 66 |
AF367998.1 | - | 2 - 3 | 336 | 66 | |
AK000780.1 | - | 2 - 3 | 336 | 66 | |
AK315031.1 | - | 2 - 3 | 275 | 66 | |
BC049210.1 | - | 2 - 3 | 300 | 66 | |
KU178816.1 | - | 2 - 3 | 229 | 66 | |
KU178817.1 | - | 2 - 3 | 229 | 66 |