Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABI3
Entrez Gene ID
51225
Gene Name
ABI family member 3
Gene Aliases
NESH, SSH3BP3
Chromosome Location
Chr.17: 49210227 - 49223225 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135186.1 NP_001128658.1 3 - 4 939 88
NM_016428.2 NP_057512.1 3 - 4 957 88
XM_005257429.2 XP_005257486.1 3 - 4 602 88
XM_011524873.1 XP_011523175.1 3 - 4 602 88
XM_017024721.1 XP_016880210.1 3 - 4 602 88
GenBank mRNA AB037886.1 - 3 - 4 957 88
AL136709.1 - 3 - 4 758 88
BC007780.2 - 3 - 4 463 88
BI768790.1 - 3 - 4 446 88

View Full Product Details