The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135186.1 | NP_001128658.1 | 5 - 6 | 1124 | 78 |
NM_016428.2 | NP_057512.1 | 5 - 6 | 1142 | 78 | |
XM_005257429.2 | XP_005257486.1 | 5 - 6 | 787 | 78 | |
GenBank mRNA | AB037886.1 | - | 5 - 6 | 1142 | 78 |
AL136709.1 | - | 5 - 6 | 943 | 78 | |
BC007780.2 | - | 5 - 6 | 648 | 78 | |
BI768790.1 | - | 5 - 6 | 631 | 78 |