The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288729.1 | NP_001275658.1 | 2 - 3 | 271 | 62 |
NM_001288730.1 | NP_001275659.1 | 2 - 3 | 261 | 62 | |
NM_001288731.1 | NP_001275660.1 | 2 - 3 | 244 | 62 | |
NM_004938.3 | NP_004929.2 | 2 - 3 | 596 | 62 | |
XM_005251757.3 | XP_005251814.1 | 2 - 3 | 366 | 62 | |
GenBank mRNA | AB208871.1 | - | 2 - 3 | 199 | 62 |
AK295119.1 | - | 2 - 3 | 328 | 62 | |
AK296839.1 | - | 2 - 3 | 541 | 62 | |
BC113660.1 | - | 1 - 2 | 134 | 62 | |
BC143730.1 | - | 1 - 2 | 133 | 62 | |
BC143733.1 | - | 1 - 2 | 134 | 62 | |
BC143759.1 | - | 1 - 2 | 134 | 62 | |
BP202018.1 | - | 2 - 3 | 271 | 62 | |
BT006935.1 | - | 1 - 2 | 61 | 62 | |
CR749834.1 | - | 2 - 3 | 244 | 62 | |
DC340223.1 | - | 2 - 3 | 261 | 62 | |
DR002874.1 | - | 2 - 3 | 554 | 62 | |
X76104.1 | - | 2 - 3 | 397 | 62 |