The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288729.1 | NP_001275658.1 | 8 - 9 | 988 | 88 |
NM_001288730.1 | NP_001275659.1 | 8 - 9 | 978 | 88 | |
NM_001288731.1 | NP_001275660.1 | 8 - 9 | 961 | 88 | |
NM_004938.3 | NP_004929.2 | 8 - 9 | 1313 | 88 | |
XM_005251757.3 | XP_005251814.1 | 8 - 9 | 1083 | 88 | |
GenBank mRNA | AB208871.1 | - | 8 - 9 | 916 | 88 |
AK295119.1 | - | 8 - 9 | 1045 | 88 | |
AK296839.1 | - | 8 - 9 | 1258 | 88 | |
BC113660.1 | - | 7 - 8 | 851 | 88 | |
BC143730.1 | - | 6 - 7 | 801 | 88 | |
BC143733.1 | - | 7 - 8 | 851 | 88 | |
BC143759.1 | - | 7 - 8 | 851 | 88 | |
BT006935.1 | - | 7 - 8 | 778 | 88 | |
CN347006.1 | - | 2 - 3 | 188 | 88 | |
CR749834.1 | - | 8 - 9 | 961 | 88 | |
X76104.1 | - | 8 - 9 | 1114 | 88 |