The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182640.2 | NP_872578.1 | 3 - 4 | 443 | 88 |
XM_011511644.2 | XP_011509946.1 | 2 - 3 | 283 | 88 | |
GenBank mRNA | AK001451.1 | - | 3 - 4 | 385 | 88 |
AK130078.1 | - | 3 - 4 | 384 | 88 | |
AV729381.1 | - | 3 - 4 | 420 | 88 | |
BC036575.1 | - | 4 - 5 | 396 | 88 | |
BC047784.1 | - | 2 - 3 | 93 | 88 | |
BC057240.1 | - | 3 - 4 | 397 | 88 | |
DT217261.1 | - | 3 - 4 | 443 | 88 |