The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153710.4 | NP_714921.4 | 1 - 2 | 145 | 112 |
NR_103997.1 | 1 - 2 | 145 | 112 | ||
GenBank mRNA | AK131533.1 | - | 1 - 2 | 105 | 112 |
AK131545.1 | - | 1 - 2 | 97 | 112 | |
AK294947.1 | - | 1 - 2 | 312 | 112 | |
BC036504.2 | - | 1 - 2 | 145 | 112 |