Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADAM11
Entrez Gene ID
4185
Gene Name
ADAM metallopeptidase domain 11
Gene Aliases
MDC
Chromosome Location
Chr.17: 44759037 - 44781846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318933.1 NP_001305862.1 12 - 13 1095 155
NM_002390.5 NP_002381.2 12 - 13 1099 155
XM_005257373.3 XP_005257430.1 12 - 13 1241 155
XM_017024647.1 XP_016880136.1 11 - 12 1013 155
GenBank mRNA AB009675.1 - 12 - 13 1078 155
AK296512.1 - 12 - 13 1176 155
D17390.1 - 9 - 10 808 155

View Full Product Details