The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318933.1 | NP_001305862.1 | 4 - 5 | 406 | 99 |
NM_002390.5 | NP_002381.2 | 4 - 5 | 406 | 99 | |
XM_005257373.3 | XP_005257430.1 | 4 - 5 | 548 | 99 | |
XM_017024647.1 | XP_016880136.1 | 3 - 4 | 324 | 99 | |
GenBank mRNA | AB009675.1 | - | 4 - 5 | 385 | 99 |
AK296512.1 | - | 4 - 5 | 487 | 99 |