The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142605.1 | NP_001136077.1 | 13 - 14 | 1437 | 71 |
NM_001258353.1 | NP_001245282.1 | 14 - 15 | 1683 | 71 | |
NM_001258354.1 | NP_001245283.1 | 14 - 15 | 1516 | 71 | |
NM_004247.3 | NP_004238.3 | 14 - 15 | 1546 | 71 | |
GenBank mRNA | AJ505017.1 | - | 10 - 11 | 920 | 71 |
AK126193.1 | - | 4 - 5 | 1013 | 71 | |
AK126464.1 | - | 15 - 16 | 1352 | 71 | |
AK128148.1 | - | 14 - 15 | 1683 | 71 | |
AK225335.1 | - | 14 - 15 | 1306 | 71 | |
AK296367.1 | - | 13 - 14 | 1240 | 71 | |
AK297392.1 | - | 14 - 15 | 1322 | 71 | |
AK302828.1 | - | 14 - 15 | 1186 | 71 | |
AK316098.1 | - | 13 - 14 | 1257 | 71 | |
BC002360.2 | - | 14 - 15 | 1316 | 71 | |
CR456774.1 | - | 13 - 14 | 1285 | 71 | |
D21163.1 | - | 14 - 15 | 1345 | 71 |