The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006039.4 | NP_006030.2 | 24 - 25 | 3979 | 88 |
GenBank mRNA | AB014609.2 | - | 24 - 25 | 3979 | 88 |
AF107292.1 | - | 24 - 25 | 3620 | 88 | |
AF134838.1 | - | 24 - 25 | 3701 | 88 | |
BC146647.1 | - | 24 - 25 | 3698 | 88 | |
BC150212.1 | - | 24 - 25 | 3683 | 88 | |
BC153884.1 | - | 24 - 25 | 3979 | 88 |