Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDZRN3
Entrez Gene ID
23024
Gene Name
PDZ domain containing ring finger 3
Gene Aliases
LNX3, SEMACAP3, SEMCAP3
Chromosome Location
Chr.3: 73382430 - 73625003 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001303139.1 NP_001290068.1 3 - 4 697 113
NM_001303140.1 NP_001290069.1 3 - 4 560 113
NM_001303141.1 NP_001290070.1 3 - 4 510 113
NM_001303142.1 NP_001290071.1 3 - 4 690 113
NM_015009.2 NP_055824.1 5 - 6 1346 113
XM_017005942.1 XP_016861431.1 4 - 5 1341 113
GenBank mRNA AB029018.1 - 5 - 6 1346 113
AK294761.1 - 3 - 4 501 113
AK299275.1 - 3 - 4 510 113
AK299560.1 - 3 - 4 544 113
AK307818.1 - 3 - 4 643 113
AK316244.1 - 3 - 4 510 113
AK316275.1 - 3 - 4 529 113
BC150623.1 - 5 - 6 1333 113
BC152417.1 - 5 - 6 1369 113

View Full Product Details