The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320126.1 | NP_001307055.1 | 7 - 8 | 1183 | 85 |
NM_020676.6 | NP_065727.4 | 6 - 7 | 1118 | 85 | |
XM_005265334.3 | XP_005265391.1 | 7 - 8 | 1179 | 85 | |
XM_005265335.3 | XP_005265392.1 | 6 - 7 | 1088 | 85 | |
XM_017006930.1 | XP_016862419.1 | 6 - 7 | 890 | 85 | |
GenBank mRNA | AF225418.1 | - | 6 - 7 | 940 | 85 |
AK122983.1 | - | 7 - 8 | 875 | 85 | |
AK172797.1 | - | 6 - 7 | 1039 | 85 | |
AK225942.1 | - | 8 - 9 | 1392 | 85 | |
AK313168.1 | - | 6 - 7 | 1009 | 85 | |
BC001698.2 | - | 6 - 7 | 791 | 85 |