The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261.3 | NP_001252.1 | 1 - 2 | 215 | 98 |
XM_017014184.1 | XP_016869673.1 | 1 - 2 | 81 | 98 | |
GenBank mRNA | AB451289.1 | - | 1 - 2 | 92 | 98 |
AB451421.1 | - | 1 - 2 | 92 | 98 | |
AK313828.1 | - | 1 - 2 | 144 | 98 | |
BC001968.1 | - | 1 - 2 | 152 | 98 | |
BT019903.1 | - | 1 - 2 | 92 | 98 | |
DB120461.1 | - | 1 - 2 | 215 | 98 | |
L25676.1 | - | 1 - 2 | 156 | 98 | |
X80230.1 | - | 1 - 2 | 190 | 98 |