The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005909.2 | NP_001005909.1 | 5 - 6 | 1031 | 85 |
NM_016291.3 | NP_057375.2 | 5 - 6 | 1034 | 85 | |
XM_006713199.3 | XP_006713262.1 | 6 - 7 | 1138 | 85 | |
XM_006713200.1 | XP_006713263.1 | 6 - 7 | 1108 | 85 | |
XM_006713201.1 | XP_006713264.1 | 6 - 7 | 1185 | 85 | |
XM_006713202.1 | XP_006713265.1 | 6 - 7 | 1187 | 85 | |
XM_011533816.1 | XP_011532118.1 | 6 - 7 | 1168 | 85 | |
XM_011533817.1 | XP_011532119.1 | 7 - 8 | 2073 | 85 | |
XM_011533818.1 | XP_011532120.1 | 6 - 7 | 2341 | 85 | |
XM_017006583.1 | XP_016862072.1 | 5 - 6 | 1030 | 85 | |
XM_017006584.1 | XP_016862073.1 | 5 - 6 | 1005 | 85 | |
XM_017006585.1 | XP_016862074.1 | 6 - 7 | 1120 | 85 | |
XM_017006586.1 | XP_016862075.1 | 6 - 7 | 2981 | 85 | |
XM_017006587.1 | XP_016862076.1 | 9 - 10 | 2330 | 85 | |
GenBank mRNA | AF177145.1 | - | 4 - 5 | 928 | 85 |
AK024045.1 | - | 5 - 6 | 971 | 85 | |
AK315204.1 | - | 5 - 6 | 972 | 85 | |
AL117458.1 | - | 3 - 4 | 558 | 85 | |
AY007091.1 | - | 5 - 6 | 957 | 85 | |
BC019694.1 | - | 5 - 6 | 943 | 85 | |
BC065533.1 | - | 5 - 6 | 960 | 85 |